Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATACTTAGCTATGAAAAGTTCCAA[C/G]CAGAATAGATAGATCAATGAAGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610174 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFCAB9 - EF-hand calcium binding domain 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBTD2 - ubiquitin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152277.2 | 1705 | UTR 3 | NP_689490.2 | |||
XM_017010022.1 | 1705 | UTR 3 | XP_016865511.1 | |||
XM_017010023.1 | 1705 | UTR 3 | XP_016865512.1 |