Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600505 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF37 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF37 - Rho guanine nucleotide exchange factor 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001669.2 | Intron | NP_001001669.2 | ||||
XM_005268447.4 | Intron | XP_005268504.1 | ||||
XM_005268448.1 | Intron | XP_005268505.1 | ||||
XM_006714784.3 | Intron | XP_006714847.1 | ||||
XM_011537642.2 | Intron | XP_011535944.1 | ||||
XM_017009460.1 | Intron | XP_016864949.1 | ||||
XM_017009461.1 | Intron | XP_016864950.1 |
CSNK1A1 - casein kinase 1 alpha 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |