Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATCATTATCCATCATTTCCTTT[A/G]CCTTCATCCTTTATACAGTCAAATC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614410 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFAP1L1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AFAP1L1 - actin filament associated protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146337.2 | Intron | NP_001139809.1 | ||||
NM_001323062.1 | Intron | NP_001309991.1 | ||||
NM_001323063.1 | Intron | NP_001309992.1 | ||||
NM_152406.3 | Intron | NP_689619.1 | ||||
XM_011537558.1 | Intron | XP_011535860.1 | ||||
XM_017009036.1 | Intron | XP_016864525.1 |