Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGATCTGAGAATCCATTCTGCCT[G/T]TGTCTTGCCTCCGTTGAACCAGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614410 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFAP1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AFAP1L1 - actin filament associated protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146337.2 | Intron | NP_001139809.1 | ||||
NM_001323062.1 | Intron | NP_001309991.1 | ||||
NM_001323063.1 | Intron | NP_001309992.1 | ||||
NM_152406.3 | Intron | NP_689619.1 | ||||
XM_011537558.1 | Intron | XP_011535860.1 | ||||
XM_017009036.1 | Intron | XP_016864525.1 |