Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGAGGGTTTTACTTAGTGAACA[C/T]GGCAGCTTGGCTGGAGGAGGGGGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614988 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPS8L2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EPS8L2 - EPS8 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022772.3 | Intron | NP_073609.2 | ||||
XM_017018130.1 | Intron | XP_016873619.1 | ||||
XM_017018131.1 | Intron | XP_016873620.1 | ||||
XM_017018132.1 | Intron | XP_016873621.1 |
Set Membership: |
HapMap Validated |