Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTAATGATGGGAGACGAAACAA[C/T]AGGGCTTTTTATTTTTTTTTAATGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM59 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TMEM59 - transmembrane protein 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305043.1 | 2654 | UTR 3 | NP_001291972.1 | |||
NM_001305049.1 | 2654 | UTR 3 | NP_001291978.1 | |||
NM_001305050.1 | 2654 | UTR 3 | NP_001291979.1 | |||
NM_001305051.1 | 2654 | UTR 3 | NP_001291980.1 | |||
NM_001305052.1 | 2654 | UTR 3 | NP_001291981.1 | |||
NM_001305066.1 | 2654 | Intron | NP_001291995.1 | |||
NM_004872.4 | 2654 | UTR 3 | NP_004863.2 | |||
XM_006711051.1 | 2654 | Intron | XP_006711114.1 |