Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGGGGTGGAGTATGTGTTGGGA[C/G]GGGGGACCTCCTGTCCAGTCCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603928 MIM: 607717 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF4B - eukaryotic translation initiation factor 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC283335 - uncharacterized LOC283335 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6757 - microRNA 6757 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNS2 - tensin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015319.2 | 1583 | Missense Mutation | ACG,AGG | T,R 10 | NP_056134.2 | |
NM_170754.2 | 1583 | Intron | NP_736610.2 | |||
NM_198316.1 | 1583 | Intron | NP_938072.1 | |||
XM_006719302.3 | 1583 | Missense Mutation | ACG,AGG | T,R 10 | XP_006719365.1 | |
XM_006719303.1 | 1583 | Intron | XP_006719366.1 | |||
XM_011538079.1 | 1583 | Intron | XP_011536381.1 | |||
XM_017019088.1 | 1583 | Intron | XP_016874577.1 | |||
XM_017019089.1 | 1583 | Intron | XP_016874578.1 |