Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616848 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIER1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR78 - WD repeat domain 78 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024763.4 | 21 | Intron | NP_079039.4 | |||
NM_207014.2 | 21 | Intron | NP_996897.2 | |||
XM_005271212.3 | 21 | Intron | XP_005271269.1 | |||
XM_011542161.2 | 21 | Intron | XP_011540463.1 | |||
XM_011542162.2 | 21 | Intron | XP_011540464.1 | |||
XM_017002353.1 | 21 | Intron | XP_016857842.1 | |||
XM_017002354.1 | 21 | Intron | XP_016857843.1 | |||
XM_017002355.1 | 21 | Intron | XP_016857844.1 | |||
XM_017002356.1 | 21 | Intron | XP_016857845.1 | |||
XM_017002357.1 | 21 | Intron | XP_016857846.1 |