Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604013 MIM: 601019 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GALT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
B4GALT2 - beta-1,4-galactosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCDC24 - coiled-coil domain containing 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC6A9 - solute carrier family 6 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024845.2 | 2341 | UTR 3 | NP_001020016.1 | |||
NM_001261380.1 | 2341 | UTR 3 | NP_001248309.1 | |||
NM_006934.3 | 2341 | UTR 3 | NP_008865.2 | |||
NM_201649.3 | 2341 | UTR 3 | NP_964012.2 | |||
XM_011542017.2 | 2341 | Intron | XP_011540319.1 | |||
XM_017002151.1 | 2341 | UTR 3 | XP_016857640.1 | |||
XM_017002152.1 | 2341 | UTR 3 | XP_016857641.1 | |||
XM_017002153.1 | 2341 | UTR 3 | XP_016857642.1 |
Set Membership: |
HapMap JSNP |