Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATCCTGCAGAAAATAGCCTAAC[A/G]AGGAATAAAAACTGACAATAACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616035 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXJ3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXJ3 - forkhead box J3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198850.1 | 2530 | UTR 3 | NP_001185779.1 | |||
NM_001198851.1 | 2530 | UTR 3 | NP_001185780.1 | |||
NM_001198852.1 | 2530 | UTR 3 | NP_001185781.1 | |||
NM_014947.4 | 2530 | UTR 3 | NP_055762.3 | |||
XM_005270632.4 | 2530 | UTR 3 | XP_005270689.1 | |||
XM_006710458.3 | 2530 | UTR 3 | XP_006710521.1 | |||
XM_006710459.3 | 2530 | UTR 3 | XP_006710522.1 | |||
XM_011541026.2 | 2530 | UTR 3 | XP_011539328.1 | |||
XM_017000693.1 | 2530 | UTR 3 | XP_016856182.1 | |||
XM_017000694.1 | 2530 | UTR 3 | XP_016856183.1 | |||
XM_017000695.1 | 2530 | UTR 3 | XP_016856184.1 |