Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616885 MIM: 601785 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARHSP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CARHSP1 - calcium regulated heat stable protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042476.2 | 1021 | UTR 3 | NP_001035941.1 | |||
NM_001278260.1 | 1021 | UTR 3 | NP_001265189.1 | |||
NM_001278261.1 | 1021 | UTR 3 | NP_001265190.1 | |||
NM_001278262.1 | 1021 | UTR 3 | NP_001265191.1 | |||
NM_001278263.1 | 1021 | UTR 3 | NP_001265192.1 | |||
NM_001278264.1 | 1021 | UTR 3 | NP_001265193.1 | |||
NM_001278265.1 | 1021 | UTR 3 | NP_001265194.1 | |||
NM_001278266.1 | 1021 | UTR 3 | NP_001265195.1 | |||
NM_014316.3 | 1021 | UTR 3 | NP_055131.2 | |||
XM_005255228.4 | 1021 | UTR 3 | XP_005255285.3 | |||
XM_005255229.4 | 1021 | UTR 3 | XP_005255286.1 | |||
XM_011522444.2 | 1021 | UTR 3 | XP_011520746.1 |
PMM2 - phosphomannomutase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |