Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGTTAAGTTAGCCACAAATACAG[C/G]GTCTGCCCACATCTTTGCTTTGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601444 MIM: 155740 MIM: 600536 MIM: 601617 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLOC1S1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BLOC1S1 - biogenesis of lysosomal organelles complex 1 subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BLOC1S1-RDH5 - BLOC1S1-RDH5 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CD63 - CD63 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITGA7 - integrin subunit alpha 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RDH5 - retinol dehydrogenase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199771.1 | Intron | NP_001186700.1 | ||||
NM_002905.3 | Intron | NP_002896.2 |
Set Membership: |
HapMap Validated |