Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611505 MIM: 603479 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CENPP - centromere protein P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012267.2 | 1132 | Intron | NP_001012267.1 | |||
NM_001286969.1 | 1132 | Intron | NP_001273898.1 | |||
NM_001286971.1 | 1132 | Intron | NP_001273900.1 | |||
XM_011518685.2 | 1132 | Intron | XP_011516987.1 | |||
XM_011518689.1 | 1132 | UTR 3 | XP_011516991.1 | |||
XM_017014715.1 | 1132 | Intron | XP_016870204.1 |
ECM2 - extracellular matrix protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197295.1 | 1132 | Intron | NP_001184224.1 | |||
NM_001197296.1 | 1132 | UTR 3 | NP_001184225.1 | |||
NM_001393.3 | 1132 | Intron | NP_001384.1 | |||
XM_017014376.1 | 1132 | Intron | XP_016869865.1 | |||
XM_017014377.1 | 1132 | Intron | XP_016869866.1 |