Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGCCGTCTTTGCAGTGGGCGATA[C/T]CTTTCTAAGGGGTTAAGTGTTATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608892 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHD7 - chromodomain helicase DNA binding protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316690.1 | Intron | NP_001303619.1 | ||||
NM_017780.3 | Intron | NP_060250.2 | ||||
XM_011517553.2 | Intron | XP_011515855.1 | ||||
XM_011517554.2 | Intron | XP_011515856.1 | ||||
XM_011517555.2 | Intron | XP_011515857.1 | ||||
XM_011517560.2 | Intron | XP_011515862.1 | ||||
XM_017013612.1 | Intron | XP_016869101.1 | ||||
XM_017013613.1 | Intron | XP_016869102.1 |