Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGGGTGGGGGTTGGTGGAGAGA[C/G]AGCGTGCGTATATTGAGTGCCTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605427 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPV4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TRPV4 - transient receptor potential cation channel subfamily V member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177428.1 | Intron | NP_001170899.1 | ||||
NM_001177431.1 | Intron | NP_001170902.1 | ||||
NM_001177433.1 | Intron | NP_001170904.1 | ||||
NM_021625.4 | Intron | NP_067638.3 | ||||
NM_147204.2 | Intron | NP_671737.1 | ||||
XM_005253918.1 | Intron | XP_005253975.1 | ||||
XM_011538630.2 | Intron | XP_011536932.2 | ||||
XM_011538631.2 | Intron | XP_011536933.2 | ||||
XM_011538632.2 | Intron | XP_011536934.2 | ||||
XM_011538633.2 | Intron | XP_011536935.2 | ||||
XM_011538634.2 | Intron | XP_011536936.2 | ||||
XM_011538635.2 | Intron | XP_011536937.1 | ||||
XM_011538636.2 | Intron | XP_011536938.1 | ||||
XM_017019774.1 | Intron | XP_016875263.1 |
Set Membership: |
Validated |