Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605715 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD276 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD276 - CD276 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024736.1 | Intron | NP_001019907.1 | ||||
NM_025240.2 | Intron | NP_079516.1 | ||||
XM_005254700.4 | Intron | XP_005254757.1 | ||||
XM_011522095.2 | Intron | XP_011520397.1 | ||||
XM_011522096.2 | Intron | XP_011520398.1 | ||||
XM_017022638.1 | Intron | XP_016878127.1 |
Set Membership: |
HapMap |