Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612768 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf45 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C4orf45 - chromosome 4 open reading frame 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152543.2 | 4640 | Intron | NP_689756.2 | |||
XM_011531671.2 | 4640 | Intron | XP_011529973.1 | |||
XM_011531672.2 | 4640 | Intron | XP_011529974.1 | |||
XM_017007808.1 | 4640 | Intron | XP_016863297.1 | |||
XM_017007809.1 | 4640 | Intron | XP_016863298.1 | |||
XM_017007810.1 | 4640 | Intron | XP_016863299.1 |
FNIP2 - folliculin interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323916.1 | 4640 | UTR 3 | NP_001310845.1 | |||
NM_020840.2 | 4640 | UTR 3 | NP_065891.1 | |||
XM_005263156.1 | 4640 | UTR 3 | XP_005263213.1 | |||
XM_005263158.1 | 4640 | UTR 3 | XP_005263215.1 | |||
XM_005263160.3 | 4640 | Intron | XP_005263217.1 | |||
XM_011532154.2 | 4640 | UTR 3 | XP_011530456.1 | |||
XM_017008487.1 | 4640 | UTR 3 | XP_016863976.1 |