Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165020 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ROS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ROS1 - ROS proto-oncogene 1, receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002944.2 | 7465 | Intron | NP_002935.2 | |||
XM_006715548.3 | 7465 | UTR 3 | XP_006715611.1 | |||
XM_011536049.2 | 7465 | UTR 3 | XP_011534351.1 | |||
XM_011536050.2 | 7465 | UTR 3 | XP_011534352.1 | |||
XM_011536051.2 | 7465 | UTR 3 | XP_011534353.1 | |||
XM_011536052.2 | 7465 | UTR 3 | XP_011534354.1 | |||
XM_011536053.2 | 7465 | UTR 3 | XP_011534355.1 | |||
XM_011536054.2 | 7465 | Intron | XP_011534356.1 | |||
XM_011536055.2 | 7465 | Intron | XP_011534357.1 | |||
XM_011536056.2 | 7465 | Intron | XP_011534358.1 | |||
XM_011536057.2 | 7465 | Intron | XP_011534359.1 | |||
XM_011536058.2 | 7465 | Intron | XP_011534360.1 | |||
XM_017011172.1 | 7465 | UTR 3 | XP_016866661.1 | |||
XM_017011173.1 | 7465 | UTR 3 | XP_016866662.1 |
Set Membership: |
HapMap Validated |