Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610652 | ||||||||||||||||||||
Literature Links: |
LINC00473 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC00473 - long intergenic non-protein coding RNA 473 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE10A - phosphodiesterase 10A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130690.2 | Intron | NP_001124162.1 | ||||
NM_006661.3 | Intron | NP_006652.1 | ||||
XM_006715321.3 | Intron | XP_006715384.1 | ||||
XM_011535387.2 | Intron | XP_011533689.2 | ||||
XM_011535388.2 | Intron | XP_011533690.1 | ||||
XM_011535393.2 | Intron | XP_011533695.1 | ||||
XM_017010194.1 | Intron | XP_016865683.1 | ||||
XM_017010195.1 | Intron | XP_016865684.1 | ||||
XM_017010196.1 | Intron | XP_016865685.1 | ||||
XM_017010197.1 | Intron | XP_016865686.1 |