Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600445 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADORA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ADORA3 - adenosine A3 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000677.3 | 1564 | Missense Mutation | AAG,ATG | K,M 266 | NP_000668.1 | |
NM_001302678.1 | 1564 | UTR 3 | NP_001289607.1 | |||
NM_001302679.1 | 1564 | Missense Mutation | AAG,ATG | K,M 121 | NP_001289608.1 |
TMIGD3 - transmembrane and immunoglobulin domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081976.2 | 1564 | Intron | NP_001075445.1 | |||
NM_001302680.1 | 1564 | Intron | NP_001289609.1 | |||
NM_020683.6 | 1564 | Intron | NP_065734.5 |
Set Membership: |
HapMap |