Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604543 MIM: 606569 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIMD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LIMD1 - LIM domains containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIMD1-AS1 - LIMD1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SACM1L - SAC1 suppressor of actin mutations 1-like (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319072.1 | Intron | NP_001306001.1 | ||||
NM_001319073.1 | Intron | NP_001306002.1 | ||||
NM_014016.4 | Intron | NP_054735.3 | ||||
XM_011533500.2 | Intron | XP_011531802.1 |
Set Membership: |
HapMap |