Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTTAAAATATGCTTTCCCCAAA[C/T]CGTTTACGTCTTTTCCATGCCTAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601175 MIM: 605964 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARL2 - ADP ribosylation factor like GTPase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ARL2-SNX15 - ARL2-SNX15 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6879 - microRNA 6879 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX15 - sorting nexin 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013306.4 | Intron | NP_037438.2 | ||||
NM_147777.3 | Intron | NP_680086.2 |
Set Membership: |
HapMap |