Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600564 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITIH4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITIH4 - inter-alpha-trypsin inhibitor heavy chain family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR8064 - microRNA 8064 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MUSTN1 - musculoskeletal, embryonic nuclear protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM110 - transmembrane protein 110 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198563.2 | 1080 | UTR 3 | NP_940965.1 |
TMEM110-MUSTN1 - TMEM110-MUSTN1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198974.2 | 1080 | Intron | NP_001185903.2 |