Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRRC2C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRRC2C - proline rich coiled-coil 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015172.3 | Intron | NP_055987.2 | ||||
XM_005245015.2 | Intron | XP_005245072.1 | ||||
XM_005245016.4 | Intron | XP_005245073.1 | ||||
XM_005245017.2 | Intron | XP_005245074.1 | ||||
XM_005245018.2 | Intron | XP_005245075.1 | ||||
XM_005245019.2 | Intron | XP_005245076.1 | ||||
XM_005245020.2 | Intron | XP_005245077.1 | ||||
XM_005245023.2 | Intron | XP_005245080.1 | ||||
XM_005245024.2 | Intron | XP_005245081.1 | ||||
XM_005245025.2 | Intron | XP_005245082.1 | ||||
XM_005245026.2 | Intron | XP_005245083.1 | ||||
XM_005245027.2 | Intron | XP_005245084.1 | ||||
XM_005245028.2 | Intron | XP_005245085.1 | ||||
XM_005245029.2 | Intron | XP_005245086.1 | ||||
XM_006711238.3 | Intron | XP_006711301.1 | ||||
XM_017000760.1 | Intron | XP_016856249.1 | ||||
XM_017000761.1 | Intron | XP_016856250.1 | ||||
XM_017000762.1 | Intron | XP_016856251.1 |