Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600979 MIM: 600228 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LTBR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LTBR - lymphotoxin beta receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270987.1 | Intron | NP_001257916.1 | ||||
NM_002342.2 | Intron | NP_002333.1 | ||||
XM_005253688.1 | Intron | XP_005253745.1 | ||||
XM_006718983.2 | Intron | XP_006719046.1 |
SCNN1A - sodium channel epithelial 1 alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038.5 | Intron | NP_001029.1 | ||||
NM_001159575.1 | Intron | NP_001153047.1 | ||||
NM_001159576.1 | Intron | NP_001153048.1 |
Set Membership: |
HapMap |