Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608862 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPATCH4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPATCH4 - G-patch domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015590.3 | 1609 | UTR 3 | NP_056405.2 | |||
NM_182679.2 | 1609 | UTR 3 | NP_872620.1 | |||
XM_005245287.3 | 1609 | UTR 3 | XP_005245344.1 |
HAPLN2 - hyaluronan and proteoglycan link protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAXE - NAD(P)HX epimerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144772.2 | 1609 | Intron | NP_658985.2 | |||
XM_017000319.1 | 1609 | Intron | XP_016855808.1 |
TTC24 - tetratricopeptide repeat domain 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |