Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTCTAACAGAGAGTAGCTGCTG[A/T]TAATGAAGAAGATAATAAAGGTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608692 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLZF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BLZF1 - basic leucine zipper nuclear factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCDC181 - coiled-coil domain containing 181 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300968.1 | Intron | NP_001287897.1 | ||||
NM_001300969.1 | Intron | NP_001287898.1 | ||||
NM_021179.2 | Intron | NP_067002.1 | ||||
XM_005245383.1 | Intron | XP_005245440.1 | ||||
XM_011509827.2 | Intron | XP_011508129.1 | ||||
XM_017001938.1 | Intron | XP_016857427.1 | ||||
XM_017001939.1 | Intron | XP_016857428.1 | ||||
XM_017001940.1 | Intron | XP_016857429.1 |