Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601476 MIM: 115437 | ||||||||||||||||||||
Literature Links: |
LAPTM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAPTM5 - lysosomal protein transmembrane 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006762.2 | 2007 | UTR 3 | NP_006753.1 | |||
XM_011542098.2 | 2007 | UTR 3 | XP_011540400.1 |
MATN1 - matrilin 1, cartilage matrix protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MATN1-AS1 - MATN1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4420 - microRNA 4420 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |