Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607645 MIM: 606046 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NSG1 - neuron specific gene family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040101.1 | 1798 | Intron | NP_001035190.1 | |||
NM_001287763.1 | 1798 | Intron | NP_001274692.1 | |||
NM_001287764.1 | 1798 | Intron | NP_001274693.1 | |||
NM_014392.4 | 1798 | Intron | NP_055207.1 | |||
XM_017008022.1 | 1798 | Intron | XP_016863511.1 |
STX18 - syntaxin 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016930.2 | 1798 | UTR 3 | NP_058626.1 | |||
XM_011513480.1 | 1798 | Intron | XP_011511782.1 | |||
XM_011513483.1 | 1798 | Intron | XP_011511785.1 | |||
XM_011513485.2 | 1798 | Intron | XP_011511787.1 | |||
XM_017008296.1 | 1798 | Intron | XP_016863785.1 | |||
XM_017008297.1 | 1798 | Intron | XP_016863786.1 | |||
XM_017008298.1 | 1798 | Intron | XP_016863787.1 |
Set Membership: |
HapMap |