Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAAGTCAAATGGACTCAGAGTG[A/G]TTCTGCCTTGTCTCTGCCTTCTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606126 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL2L14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCL2L14 - BCL2 like 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030766.1 | Intron | NP_110393.1 | ||||
NM_138722.1 | Intron | NP_620048.1 | ||||
NM_138723.1 | Intron | NP_620049.1 | ||||
XM_006719151.3 | Intron | XP_006719214.1 | ||||
XM_011520848.1 | Intron | XP_011519150.1 | ||||
XM_011520849.1 | Intron | XP_011519151.1 | ||||
XM_017019963.1 | Intron | XP_016875452.1 |