Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606832 MIM: 609497 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ERAP1 - endoplasmic reticulum aminopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040458.1 | Intron | NP_001035548.1 | ||||
NM_001198541.1 | Intron | NP_001185470.1 | ||||
NM_016442.3 | Intron | NP_057526.3 | ||||
XM_005272015.4 | Intron | XP_005272072.1 | ||||
XM_005272016.4 | Intron | XP_005272073.1 | ||||
XM_011543480.2 | Intron | XP_011541782.1 | ||||
XM_011543481.2 | Intron | XP_011541783.1 | ||||
XM_011543484.2 | Intron | XP_011541786.1 | ||||
XM_011543485.2 | Intron | XP_011541787.1 | ||||
XM_011543486.2 | Intron | XP_011541788.1 | ||||
XM_017009580.1 | Intron | XP_016865069.1 | ||||
XM_017009581.1 | Intron | XP_016865070.1 | ||||
XM_017009582.1 | Intron | XP_016865071.1 | ||||
XM_017009583.1 | Intron | XP_016865072.1 |
ERAP2 - endoplasmic reticulum aminopeptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011543544.1 | Intron | XP_011541846.1 |
Set Membership: |
HapMap |