Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTAAATGAGAAAACAGTTCATCA[A/G]TGGATTTATAAGGTAATATCATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154200 MIM: 613580 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MDH1 - malate dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199111.1 | Intron | NP_001186040.1 | ||||
NM_001199112.1 | Intron | NP_001186041.1 | ||||
NM_001316374.1 | Intron | NP_001303303.1 | ||||
NM_005917.3 | Intron | NP_005908.1 |
WDPCP - WD repeat containing planar cell polarity effector | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042692.2 | Intron | NP_001036157.1 | ||||
NM_015910.5 | Intron | NP_056994.3 | ||||
XM_005264348.3 | Intron | XP_005264405.1 | ||||
XM_011532881.2 | Intron | XP_011531183.1 | ||||
XM_011532884.2 | Intron | XP_011531186.1 | ||||
XM_011532886.2 | Intron | XP_011531188.1 | ||||
XM_011532887.2 | Intron | XP_011531189.1 | ||||
XM_011532890.2 | Intron | XP_011531192.1 | ||||
XM_011532891.2 | Intron | XP_011531193.1 | ||||
XM_017004251.1 | Intron | XP_016859740.1 | ||||
XM_017004252.1 | Intron | XP_016859741.1 | ||||
XM_017004253.1 | Intron | XP_016859742.1 | ||||
XM_017004254.1 | Intron | XP_016859743.1 |
Set Membership: |
HapMap |