Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGCATCATAGATGTAGAACATT[G/T]GAGAATTTTGAAAGTTCTTCAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603076 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABCG1 - ATP binding cassette subfamily G member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004915.3 | Intron | NP_004906.3 | ||||
NM_016818.2 | Intron | NP_058198.2 | ||||
NM_207174.1 | Intron | NP_997057.1 | ||||
NM_207627.1 | Intron | NP_997510.1 | ||||
NM_207628.1 | Intron | NP_997511.1 | ||||
NM_207629.1 | Intron | NP_997512.1 | ||||
XM_011529806.1 | Intron | XP_011528108.1 | ||||
XM_011529807.2 | Intron | XP_011528109.1 |
Set Membership: |
HapMap |