Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125265 MIM: 182175 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
REEP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
REEP5 - receptor accessory protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005669.4 | 1392 | UTR 3 | NP_005660.4 | |||
XM_017009844.1 | 1392 | Intron | XP_016865333.1 |
SRP19 - signal recognition particle 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204193.1 | 1392 | Intron | NP_001191122.1 | |||
NM_001204194.1 | 1392 | Intron | NP_001191123.1 | |||
NM_001204196.1 | 1392 | Intron | NP_001191125.1 | |||
NM_001204199.1 | 1392 | Intron | NP_001191128.1 | |||
NM_003135.2 | 1392 | Intron | NP_003126.1 |
Set Membership: |
HapMap |