Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTTTTGTATAAACCCTAATAAC[C/T]GTAGTAGGTAACTTTACAGAGTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604731 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6125 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MIR6125 - microRNA 6125 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP15 - ubiquitin specific peptidase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252078.1 | Intron | NP_001239007.1 | ||||
NM_001252079.1 | Intron | NP_001239008.1 | ||||
NM_006313.2 | Intron | NP_006304.1 | ||||
XM_005269261.2 | Intron | XP_005269318.1 | ||||
XM_006719718.3 | Intron | XP_006719781.1 | ||||
XM_017020284.1 | Intron | XP_016875773.1 | ||||
XM_017020285.1 | Intron | XP_016875774.1 |
Set Membership: |
HapMap Validated |