Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGCAGCCTTCTGGTCAGCACTC[A/C]TAGGCCAGCACAGGCCCTGGGTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600702 | ||||||||||||||||||||
Literature Links: |
FIGNL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FIGNL2 - fidgetin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013690.4 | 4251 | UTR 3 | NP_001013712.4 | |||
XM_005268870.3 | 4251 | UTR 3 | XP_005268927.1 | |||
XM_017019298.1 | 4251 | UTR 3 | XP_016874787.1 | |||
XM_017019299.1 | 4251 | UTR 3 | XP_016874788.1 |
SCN8A - sodium voltage-gated channel alpha subunit 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |