Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600050 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAP3K11 - mitogen-activated protein kinase kinase kinase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002419.3 | 666 | Intron | NP_002410.1 |
PCNX3 - pecanex homolog 3 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032223.3 | 666 | Intron | NP_115599.2 | |||
XM_005273987.4 | 666 | UTR 5 | XP_005274044.1 | |||
XM_005273990.4 | 666 | UTR 5 | XP_005274047.1 | |||
XM_006718549.3 | 666 | UTR 5 | XP_006718612.1 | |||
XM_006718551.3 | 666 | UTR 5 | XP_006718614.1 | |||
XM_011545024.2 | 666 | Intron | XP_011543326.1 | |||
XM_011545025.2 | 666 | UTR 5 | XP_011543327.1 | |||
XM_011545026.2 | 666 | Intron | XP_011543328.1 | |||
XM_011545027.1 | 666 | Intron | XP_011543329.1 | |||
XM_011545028.2 | 666 | UTR 5 | XP_011543330.1 |
Set Membership: |
HapMap |