Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
59 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604950 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHTF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PHTF1 - putative homeodomain transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323041.1 | 2803 | Intron | NP_001309970.1 | |||
NM_001323042.1 | 2803 | Intron | NP_001309971.1 | |||
NM_001323043.1 | 2803 | Intron | NP_001309972.1 | |||
NM_001323044.1 | 2803 | Intron | NP_001309973.1 | |||
NM_001323045.1 | 2803 | Intron | NP_001309974.1 | |||
NM_001323046.1 | 2803 | Intron | NP_001309975.1 | |||
NM_001323047.1 | 2803 | UTR 3 | NP_001309976.1 | |||
NM_001323048.1 | 2803 | UTR 3 | NP_001309977.1 | |||
NM_001323049.1 | 2803 | UTR 3 | NP_001309978.1 | |||
NM_001323050.1 | 2803 | UTR 3 | NP_001309979.1 | |||
NM_001323051.1 | 2803 | Intron | NP_001309980.1 | |||
NM_001323052.1 | 2803 | Intron | NP_001309981.1 | |||
NM_001323053.1 | 2803 | Intron | NP_001309982.1 | |||
NM_006608.2 | 2803 | Intron | NP_006599.2 |
Set Membership: |
HapMap |