Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614126 MIM: 614127 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MARC1 - mitochondrial amidoxime reducing component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022746.3 | Intron | NP_073583.3 | ||||
XM_011509900.2 | Intron | XP_011508202.1 | ||||
XM_011509903.2 | Intron | XP_011508205.1 | ||||
XM_011509904.2 | Intron | XP_011508206.1 | ||||
XM_017002096.1 | Intron | XP_016857585.1 | ||||
XM_017002097.1 | Intron | XP_016857586.1 |
MARC2 - mitochondrial amidoxime reducing component 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |