Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606511 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MARK1 - microtubule affinity regulating kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286124.1 | Intron | NP_001273053.1 | ||||
NM_001286126.1 | Intron | NP_001273055.1 | ||||
NM_001286128.1 | Intron | NP_001273057.1 | ||||
NM_001286129.1 | Intron | NP_001273058.1 | ||||
NM_018650.4 | Intron | NP_061120.3 | ||||
XM_005273134.4 | Intron | XP_005273191.1 | ||||
XM_006711326.3 | Intron | XP_006711389.1 | ||||
XM_011509561.2 | Intron | XP_011507863.1 | ||||
XM_011509562.2 | Intron | XP_011507864.1 | ||||
XM_017001305.1 | Intron | XP_016856794.1 | ||||
XM_017001306.1 | Intron | XP_016856795.1 | ||||
XM_017001307.1 | Intron | XP_016856796.1 |
Set Membership: |
HapMap |