Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCAAGCACAAAGAGGAGAAGGC[C/T]GACCTGGACCCCAGTAAGTTGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142970 MIM: 142971 MIM: 609687 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXC-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HOXC-AS1 - HOXC cluster antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXC-AS2 - HOXC cluster antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXC8 - homeobox C8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXC9 - homeobox C9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107984512 - uncharacterized LOC107984512 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR196A2 - microRNA 196a-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |