Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611680 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf116 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C1orf116 - chromosome 1 open reading frame 116 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083924.1 | 4965 | UTR 3 | NP_001077393.1 | |||
NM_023938.5 | 4965 | UTR 3 | NP_076427.2 | |||
XM_005273259.1 | 4965 | UTR 3 | XP_005273316.1 | |||
XM_006711530.1 | 4965 | UTR 3 | XP_006711593.1 | |||
XM_011509973.2 | 4965 | UTR 3 | XP_011508275.1 |