Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147460 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SOD2 - superoxide dismutase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000636.3 | 4059 | UTR 3 | NP_000627.2 | |||
NM_001024465.2 | 4059 | UTR 3 | NP_001019636.1 | |||
NM_001024466.2 | 4059 | UTR 3 | NP_001019637.1 | |||
NM_001322814.1 | 4059 | UTR 3 | NP_001309743.1 | |||
NM_001322815.1 | 4059 | UTR 3 | NP_001309744.1 | |||
NM_001322816.1 | 4059 | Intron | NP_001309745.1 | |||
NM_001322817.1 | 4059 | UTR 3 | NP_001309746.1 | |||
NM_001322819.1 | 4059 | UTR 3 | NP_001309748.1 | |||
NM_001322820.1 | 4059 | UTR 3 | NP_001309749.1 |
Set Membership: |
HapMap Validated |