Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603473 MIM: 608141 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LATS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LATS1 - large tumor suppressor kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC645967 - uncharacterized LOC645967 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP43 - nucleoporin 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198887.2 | 3106 | UTR 3 | NP_942590.1 | |||
XM_005266960.4 | 3106 | UTR 3 | XP_005267017.1 | |||
XM_005266961.3 | 3106 | UTR 3 | XP_005267018.1 | |||
XM_005266962.3 | 3106 | UTR 3 | XP_005267019.1 | |||
XM_011535798.2 | 3106 | Intron | XP_011534100.1 | |||
XM_011535799.2 | 3106 | UTR 3 | XP_011534101.1 |