Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603184 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK8 - cyclin dependent kinase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260.2 | Intron | NP_001251.1 | ||||
NM_001318368.1 | Intron | NP_001305297.1 | ||||
XM_005266216.4 | Intron | XP_005266273.1 | ||||
XM_011534864.1 | Intron | XP_011533166.1 | ||||
XM_011534865.2 | Intron | XP_011533167.1 | ||||
XM_017020317.1 | Intron | XP_016875806.1 | ||||
XM_017020318.1 | Intron | XP_016875807.1 |