Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGCATCAACCTGGCACTGGCCT[A/G]GAGCGGCCAGGAGGCCATCTGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609701 | ||||||||||||||||||||
Literature Links: |
NAGLU PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAGLU - N-acetyl-alpha-glucosaminidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000263.3 | 580 | Nonsense Mutation | TAG,TGG | *,W 168 | NP_000254.2 | |
XM_006721920.2 | 580 | UTR 5 | XP_006721983.1 | |||
XM_011524840.1 | 580 | UTR 5 | XP_011523142.1 | |||
XM_017024686.1 | 580 | Intron | XP_016880175.1 | |||
XM_017024687.1 | 580 | UTR 5 | XP_016880176.1 |