Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604025 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AXIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AXIN2 - axin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004655.3 | 4044 | UTR 3 | NP_004646.3 | |||
XM_011525319.2 | 4044 | UTR 3 | XP_011523621.1 | |||
XM_011525320.1 | 4044 | UTR 3 | XP_011523622.1 | |||
XM_011525321.2 | 4044 | UTR 3 | XP_011523623.1 | |||
XM_017025192.1 | 4044 | UTR 3 | XP_016880681.1 | |||
XM_017025193.1 | 4044 | UTR 3 | XP_016880682.1 | |||
XM_017025194.1 | 4044 | UTR 3 | XP_016880683.1 |
Set Membership: |
HapMap Validated |