Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGAAAAATAAAAAATAAAAGAAG[A/T]CCTACCTCAGTCTCTATGATTTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133530 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BIVM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BIVM - basic, immunoglobulin-like variable motif containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BIVM-ERCC5 - BIVM-ERCC5 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204425.1 | Intron | NP_001191354.1 |
ERCC5 - ERCC excision repair 5, endonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000123.3 | Intron | NP_000114.2 |
Set Membership: |
HapMap |