Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611470 MIM: 601585 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGSN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LGSN - lengsin, lens protein with glutamine synthetase domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143940.1 | Intron | NP_001137412.1 | ||||
NM_016571.2 | Intron | NP_057655.2 | ||||
XM_011535889.2 | Intron | XP_011534191.1 | ||||
XM_011535892.2 | Intron | XP_011534194.1 | ||||
XM_017010929.1 | Intron | XP_016866418.1 | ||||
XM_017010930.1 | Intron | XP_016866419.1 | ||||
XM_017010931.1 | Intron | XP_016866420.1 |
PTP4A1 - protein tyrosine phosphatase type IVA, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003463.4 | Intron | NP_003454.1 | ||||
XM_011536111.1 | Intron | XP_011534413.1 | ||||
XM_011536112.1 | Intron | XP_011534414.1 | ||||
XM_017011270.1 | Intron | XP_016866759.1 | ||||
XM_017011271.1 | Intron | XP_016866760.1 | ||||
XM_017011272.1 | Intron | XP_016866761.1 |
Set Membership: |
HapMap |