Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609603 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYGN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CRYGN - crystallin gamma N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308292.1 | 1043 | Intron | NP_001295221.1 | |||
NM_144727.2 | 1043 | Silent Mutation | GAC,GAT | D,D 137 | NP_653328.1 | |
XM_005249952.3 | 1043 | Silent Mutation | GAC,GAT | D,D 137 | XP_005250009.1 | |
XM_011515836.1 | 1043 | Silent Mutation | GAC,GAT | D,D 137 | XP_011514138.1 | |
XM_011515837.2 | 1043 | Silent Mutation | GAC,GAT | D,D 102 | XP_011514139.1 | |
XM_017011773.1 | 1043 | Intron | XP_016867262.1 |
MIR3907 - microRNA 3907 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |